Thermo Scientific Chemicals

Heneicosanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 204690010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Heneicosanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 204690010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 204690010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2363-71-5
IUPAC Name
henicosanoic acid
Molecular Formula
C21H42O2
InChI Key
CKDDRHZIAZRDBW-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCC(O)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Appearance (Form)
Glistening crystalline powder
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text