Heneicosanoic acid, 99%, Thermo Scientific Chemicals
Heneicosanoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Heneicosanoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Catalog number 204691000
Price (USD)/ Each
-
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2363-71-5
IUPAC Namehenicosanoic acid
Molecular FormulaC21H42O2
InChI KeyCKDDRHZIAZRDBW-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Glistening crystalline powder
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only