Thermo Scientific™

AminoLink™ Plus Micro Immobilization Kit, 0.1 mL

Catalog number: 20475
Have Questions?
Thermo Scientific™

AminoLink™ Plus Micro Immobilization Kit, 0.1 mL

Catalog number: 20475
Have Questions?
Catalog Number
20475
Unit Size
10 columns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2047510 columns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific AminoLink Plus Micro Immobilization Kit enables covalent attachment of amine-containing ligands for use in antibody purification and other affinity protocols.

Features of the AminoLink Plus Micro Immobilization Kit:

Optimized for small scale—ideal for coupling small amounts (25 to 250 μg) of purified antibodies, proteins and other amine-containing ligands for purification of specific antibodies from crude serum
High-performance affinity resin—covalent attachment of the protein to AminoLink Plus Coupling Resin results in a leach-resistant affinity support
Efficient coupling chemistry—immobilization efficiency greater than 80% (as measured with 4 hour coupling reaction using mammalian antibodies)
Fast and easy to use—perform wash and elution steps using a microcentrifuge
Reusable—use prepared peptide resin several times with no significant loss of capacity

The AminoLink Plus Micro Immobilization Kit is for immobilizing small amounts (25 to 100 μg) of purified antibody and other proteins onto Thermo Scientific AminoLink Plus Coupling Resin (crosslinked 4% beaded agarose) to create a small, reusable, microcentrifuge affinity column. The coupling and affinity purification procedures are optimized for small sample volumes (200 to 300 μL). Wash and elution steps are achieved rapidly and efficiently with the convenient microcentrifuge spin columns. Each kit contains sufficient reagents for 10 coupling reactions and 20 affinity purifications.

Related Products
AminoLink™ Plus Coupling Resin
AminoLink™ Plus Immobilization Kit, 2 mL
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity (Metric)
25 to 100 μg Protein Immobilization per Column
Formulation
Multi-component kit with 10 x 0.1mL resin columns
Product Line
AminoLink™
Quantity
10 Column Kit
Target
Antibody, Proteins through Primary Amine
Type
Immobilization Kit
Stationary Phase
Aldehyde
Sufficient For
25 to 100 μg of Protein per Column
Format
Kit, Microcentrifuge, Spin Column
Column Type
Affinity, Agarose Resin

Contents & Storage

Sufficient For: Immobilizing 25 to 100 μg of protein per column; Includes buffers for 20 affinity purifications
• AminoLink Plus Spin Columns, 0.1 mL, 10 columns
• PBS Pack (makes 500 mL), 1 pack
• Sodium Cyanoborohydride Solution, 5M, 0.5 mL
• Quenching Buffer, 60 mL
• Wash Solution, 25 mL
• Elution Buffer, 50 mL
• Collection Tubes, 200 tubes

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text