Thermo Scientific Chemicals

L-alpha-Hydroxyisocaproic acid, 98%, Thermo Scientific Chemicals

Catalog number: 204810010
1 g, Each
Thermo Scientific Chemicals

L-alpha-Hydroxyisocaproic acid, 98%, Thermo Scientific Chemicals

Catalog number: 204810010
1 g, Each
Quantity
Catalog number: 204810010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13748-90-8
IUPAC Name
(2S)-2-hydroxy-4-methylpentanoic acid
Molecular Formula
C6H12O3
InChI Key
LVRFTAZAXQPQHI-YFKPBYRVSA-N
SMILES
CC(C)C[C@H](O)C(O)=O
Appearance (Color)
White to off-white
Melting point
77°C to 81°C
Titration with NaOH
>=97.5 %
Appearance (Form)
Crystalline powder
NMR
consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text