Hydroxy Naphthol Blue, disodium salt, ACS reagent
Hydroxy Naphthol Blue, disodium salt, ACS reagent
Hydroxy Naphthol Blue, disodium salt, ACS reagent
Hydroxy Naphthol Blue, disodium salt, ACS reagent
Thermo Scientific Chemicals

Hydroxy Naphthol Blue, disodium salt, ACS reagent

CAS: 165660-27-5 | C20H12N2Na2O11S3 | 600.495 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 204880050
Price (USD)
27.65
Online Exclusive
30.50
Save 2.85 (9%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
27.65
Online Exclusive
30.50
Save 2.85 (9%)
Each
Add to cart
Chemical Identifiers
CAS165660-27-5
IUPAC Name3-hydroxy-4-[2-(2-oxo-4-sulfo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]naphthalene-2,7-disulfonic acid disodium
Molecular FormulaC20H14N2Na2O11S3
InChI KeyLXJHFXHZWYBBKJ-UHFFFAOYSA-N
SMILES[Na].[Na].OC1=C(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)C2=CC=C(C=C2C=C1S(O)(=O)=O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Suitability(for Ca determination) Passes test
Appearance (Color)Blue to dark purple
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link