Chemical Identifiers
CAS165660-27-5
IUPAC Name3-hydroxy-4-[2-(2-oxo-4-sulfo-1,2-dihydronaphthalen-1-ylidene)hydrazin-1-yl]naphthalene-2,7-disulfonic acid disodium
Molecular FormulaC20H14N2Na2O11S3
InChI KeyLXJHFXHZWYBBKJ-UHFFFAOYSA-N
SMILES[Na].[Na].OC1=C(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)C2=CC=C(C=C2C=C1S(O)(=O)=O)S(O)(=O)=O
View more