Thermo Scientific Chemicals

cis-4-Hydroxy-D-proline, 99%

Catalog number: 204910100
10 g, Glass Bottle, Each
Thermo Scientific Chemicals

cis-4-Hydroxy-D-proline, 99%

Catalog number: 204910100
10 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 204910100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2584-71-6
IUPAC Name
(2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid
Molecular Formula
C5H9NO3
InChI Key
PMMYEEVYMWASQN-QWWZWVQMSA-N
SMILES
O[C@H]1CN[C@H](C1)C(O)=O
Appearance (Color)
White
Specific optical rotation
+56° to +60° (24°C, 589 nm) (c=2, water)
Appearance (Form)
Powder
Infrared spectrum
Conforms
TLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text