Thermo Scientific Chemicals

Iminodiacetic acid, 98+%

Catalog number: 204970250
25 g, Each
Thermo Scientific Chemicals

Iminodiacetic acid, 98+%

Catalog number: 204970250
25 g, Each
Quantity
Catalog number: 204970250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-73-4
IUPAC Name
2-[(carboxymethyl)amino]acetic acid
Molecular Formula
C4H7NO4
InChI Key
NBZBKCUXIYYUSX-UHFFFAOYSA-N
SMILES
OC(=O)CNCC(O)=O
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text