Thermo Scientific Chemicals

DL-Isocitric acid, trisodium salt hydrate, 95%, Thermo Scientific Chemicals

Catalog number: 205010250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-Isocitric acid, trisodium salt hydrate, 95%, Thermo Scientific Chemicals

Catalog number: 205010250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 205010250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1637-73-6
IUPAC Name
trisodium 1-hydroxypropane-1,2,3-tricarboxylate
Molecular Formula
C6H5Na3O7
InChI Key
HWMVXEKEEAIYGB-UHFFFAOYNA-K
SMILES
[Na+].[Na+].[Na+].OC(C(CC([O-])=O)C([O-])=O)C([O-])=O
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Water
=<20 % (K.F.)
Appearance (Form)
Crystalline powder
Titration with HClO4
>=94 % (On dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text