Sodium bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals
Sodium bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 205130050
Price (USD)/ Each
423.65
Online exclusive
470.00 
Save 46.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
423.65
Online exclusive
470.00 
Save 46.35 (10%)
Add to cart
Sodium bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals
Catalog number205130050
Price (USD)/ Each
423.65
Online exclusive
470.00 
Save 46.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7647-15-6
IUPAC Namesodium bromide
Molecular FormulaBrNa
InChI KeyJHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES[Na+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Argentometric>=99.0 % (on dry substance)
Heavy metals (as Pb)=<10 ppm
Loss on drying=<1.0 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only