Search Thermo Fisher Scientific
Catalog Number | Color | Diameter (Metric) |
---|---|---|
20515 | Orange | 12 mm |
20518 | Blue | 13 mm |
Cryo Bio System Round Visotubes are ideal for freezing and storage of straws containing biological materials.
Cryo Bio System High Security straws are inserted in visotubes available in polygonal, round and hexagonal shapes.
Share catalog number, name or link