Aluminum isopropoxide, 98%
Aluminum isopropoxide, 98%
Aluminum isopropoxide, 98%
Aluminum isopropoxide, 98%
Thermo Scientific Chemicals

Aluminum isopropoxide, 98%

CAS: 555-31-7 | C9H21AlO3 | 204.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 205170010
Price (USD)
78.40
Each
Estimated availability date  Pending  
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
78.40
Each
Add to cart
Chemical Identifiers
CAS555-31-7
IUPAC Namealuminium(3+) tris(propan-2-olate)
Molecular FormulaC9H21AlO3
InChI KeySMZOGRDCAXLAAR-UHFFFAOYSA-N
SMILES[Al+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Aluminium (Al)12.9 to 13.5 %
Appearance (Form)Powder
Melting point129°C to 136°C
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0460696Certificate of AnalysisMay 17, 2025205170051, 205175000, 205170010
A0469186Certificate of AnalysisJan 31, 2025205170051, 205175000, 205170010
A0466538Certificate of AnalysisOct 01, 2024205170051, 205175000, 205170010
A0466536Certificate of AnalysisOct 01, 2024205170051, 205175000, 205170010
A0466537Certificate of AnalysisOct 01, 2024205170051, 205175000, 205170010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link