Aluminum isopropoxide, 98%, Thermo Scientific Chemicals
Aluminum isopropoxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum isopropoxide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 205170051
Price (USD)/ Each
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS555-31-7
IUPAC Namealuminium(3+) tris(propan-2-olate)
Molecular FormulaC9H21AlO3
InChI KeySMZOGRDCAXLAAR-UHFFFAOYSA-N
SMILES[Al+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Aluminium (Al)12.9 to 13.5 %
Appearance (Form)Powder
Melting point129°C to 136°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only