Thermo Scientific Chemicals

Aluminum isopropoxide, 98%, Thermo Scientific Chemicals

Catalog number: 205175000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Aluminum isopropoxide, 98%, Thermo Scientific Chemicals

Catalog number: 205175000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 205175000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
555-31-7
IUPAC Name
aluminium(3+) tris(propan-2-olate)
Molecular Formula
C9H21AlO3
InChI Key
SMZOGRDCAXLAAR-UHFFFAOYSA-N
SMILES
[Al+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Appearance (Color)
White
Infrared spectrum
Conforms
Aluminium (Al)
12.9 to 13.5 %
Appearance (Form)
Powder
Melting point
129°C to 136°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text