Sodium bismuthate(V), 85%, Thermo Scientific Chemicals
Sodium bismuthate(V), 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium bismuthate(V), 85%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
Packaging:
Glass bottle
Catalog number 205210010
Price (USD)/ Each
493.00
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
493.00
Add to cart
Sodium bismuthate(V), 85%, Thermo Scientific Chemicals
Catalog number205210010
Price (USD)/ Each
493.00
-
Add to cart
Chemical Identifiers
CAS12232-99-4
IUPAC Namesodium bismuthanoylolate
Molecular FormulaBiNaO3
InChI KeyPNYYBUOBTVHFDN-UHFFFAOYSA-N
SMILES[Na+].[O-][Bi](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Titration with KMnO4>=80 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only