Thermo Scientific Chemicals

Kojic acid, 99%, Thermo Scientific Chemicals

Catalog number: 205220250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Kojic acid, 99%, Thermo Scientific Chemicals

Catalog number: 205220250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 205220250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
501-30-4
IUPAC Name
5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
Molecular Formula
C6H6O4
InChI Key
BEJNERDRQOWKJM-UHFFFAOYSA-N
SMILES
OCC1=CC(=O)C(O)=CO1
Appearance (Form)
Crystalline powder
Melting point
151°C to 156°C
Infrared spectrum
Conforms
Appearance (Color)
White to beige to light brown
Titration with NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text