Kojic acid, 99%, Thermo Scientific Chemicals
Kojic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kojic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 205221000
Price (USD)/ Each
540.65
Online exclusive
600.00 
Save 59.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
540.65
Online exclusive
600.00 
Save 59.35 (10%)
Add to cart
Kojic acid, 99%, Thermo Scientific Chemicals
Catalog number205221000
Price (USD)/ Each
540.65
Online exclusive
600.00 
Save 59.35 (10%)
-
Add to cart
Chemical Identifiers
CAS501-30-4
IUPAC Name5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
Molecular FormulaC6H6O4
InChI KeyBEJNERDRQOWKJM-UHFFFAOYSA-N
SMILESOCC1=CC(=O)C(O)=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point151°C to 156°C
Infrared spectrumConforms
Appearance (Color)White to beige to light brown
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only