Thermo Scientific Chemicals

(R)-(+)-alpha-Methylbenzyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: 205320050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

(R)-(+)-alpha-Methylbenzyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: 205320050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 205320050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
33375-06-3
Appearance (Form)
Liquid
GC
>=97.5 %
Specific optical rotation
+9° to +11° (20°C, 589 nm) (neat)
Refractive index
1.5110 to 1.5150 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text