L-Phenylalanine ethyl ester hydrochloride, 99%
L-Phenylalanine ethyl ester hydrochloride, 99%
Thermo Scientific Chemicals

L-Phenylalanine ethyl ester hydrochloride, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Catalog number 205430100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS3182-93-2
IUPAC Namehydrogen ethyl (2S)-2-amino-3-phenylpropanoate chloride
Molecular FormulaC11H16ClNO2
InChI KeyFPFQPLFYTKMCHN-PPHPATTJSA-N
SMILES[H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or needles
Appearance (Color)White
Melting point154°C to 157°C
Titration with HClO4>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only