Thermo Scientific Chemicals

Florisil, 60-100 mesh, for column chromatography, Thermo Scientific Chemicals

Catalog number: 205450010
1 kg, Glass Bottle, Each
Thermo Scientific Chemicals

Florisil, 60-100 mesh, for column chromatography, Thermo Scientific Chemicals

Catalog number: 205450010
1 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 205450010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1343-88-0
IUPAC Name
silicon(4+) magnesium(2+) trioxidandiide
Molecular Formula
MgO3Si
InChI Key
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Molecular Weight (g/mol)
100.39
Appearance (Form)
Powder
Particle size
41.0 % (80 mesh) typical
Particle size
20.0 % (70 mesh) typical
Particle size
6.9 % (140 mesh) typical
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text