Florisil™, 60-100 mesh, for column chromatography
Florisil™, 60-100 mesh, for column chromatography
Florisil™, 60-100 mesh, for column chromatography
Florisil™, 60-100 mesh, for column chromatography
Thermo Scientific Chemicals

Florisil™, 60-100 mesh, for column chromatography

Florisil, MgO3Si, CAS Number-1343-88-0, 1kg, White, 01,393; 09,223, 100.39, ZADYMNAVLSWLEQ-UHFFFAOYSA-N, 10, 5509, MFCD00078249, 60-100 mesh, for column chromatography, Glass bottle | CAS: 1343-88-0 | MgO3Si | 100.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
View all
Glass Bottle
Catalog number 205452500
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
-
Add to cart
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1343-88-0
IUPAC Namesilicon(4+) magnesium(2+) trioxidandiide
Molecular FormulaMgO3Si
InChI KeyZADYMNAVLSWLEQ-UHFFFAOYSA-N
Molecular Weight (g/mol)100.39
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatography
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468278Certificate of AnalysisJan 31, 2025205455000, 205452500, 205450010, 205450050
A0466595Certificate of AnalysisOct 10, 2024205455000, 205452500, 205450010, 205450050
A0463963Certificate of AnalysisJun 20, 2024205455000, 205452500, 205450010, 205450050
A0454505Certificate of AnalysisAug 25, 2023205455000, 205452500, 205450010, 205450050
A0423668Certificate of AnalysisApr 24, 2023205450010, 205450050, 205452500, 205455000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection