Thermo Scientific Chemicals

Manganese(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 205660025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Manganese(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 205660025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 205660025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6156-78-1
IUPAC Name
manganese(2+) diacetate tetrahydrate
Molecular Formula
C4H14MnO8
InChI Key
CESXSDZNZGSWSP-UHFFFAOYSA-L
SMILES
O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Appearance (Color)
Light pink
Titration Complexometric
>=99.0 %
Chloride (Cl)
=<20 ppm
Copper (Cu)
=<5 ppm
Insoluble matter
=<0.01 % (in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text