Mercury(II) iodide, red, 99+%, Thermo Scientific Chemicals
Mercury(II) iodide, red, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercury(II) iodide, red, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 205745000
Price (USD)/ Each
562.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
662.00 
Save 99.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
562.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
662.00 
Save 99.35 (15%)
Add to cart
Mercury(II) iodide, red, 99+%, Thermo Scientific Chemicals
Catalog number205745000
Price (USD)/ Each
562.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
662.00 
Save 99.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7774-29-0
IUPAC Namemercury(2+) diiodide
Molecular FormulaHgI2
InChI KeyYFDLHELOZYVNJE-UHFFFAOYSA-L
SMILES[I-].[I-].[Hg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Soluble Hg salts (as Hg)=<0.05 %
Appearance (Color)Red
Titration Iodometric>=99.0 %
Mercurous Mercury (as Hg)=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only