Thermo Scientific Chemicals

Silver(I) oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 205800500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Silver(I) oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 205800500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 205800500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20667-12-3
IUPAC Name
(argentiooxy)silver
Molecular Formula
Ag2O
InChI Key
KHJDQHIZCZTCAE-UHFFFAOYSA-N
SMILES
[Ag]O[Ag]
Elemental Analysis
>=99.0 %
Appearance (Form)
Powder
Aluminium (Al)
=<10 ppm
Nitrate (NO3)
=<50 ppm
Iron (Fe)
=<30 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text