Ammonium sulfate, 99.5%, for analysis
Ammonium sulfate, 99.5%, for analysis
Ammonium sulfate, 99.5%, for analysis
Ammonium sulfate, 99.5%, for analysis
Thermo Scientific Chemicals

Ammonium sulfate, 99.5%, for analysis

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 205870050
Price (USD)
363.00
Each
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
363.00
Each
Add to cart
Chemical Identifiers
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Titration excess NaOH>=99.5 %
Iron (Fe)=<5 ppm
Appearance (Form)Crystals
Residue after ignition=<0.01 % (as SO4)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection