Thermo Scientific Chemicals

Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 205880500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 205880500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 205880500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-83-7
IUPAC Name
iron(3+) ammonium dodecahydrate disulfate
Molecular Formula
FeH28NO20S2
InChI Key
LCPUDZUWZDSKMX-UHFFFAOYSA-K
SMILES
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Phosphate (PO4)
=<20 ppm
Potassium (K)
=<50 ppm
Chloride (Cl)
=<5 ppm
Zinc (Zn)
=<10 ppm
Titration Iodometric
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text