Manganese(II) chloride tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Manganese(II) chloride tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) chloride tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 205890025
Price (USD)/ Each
243.65
Online exclusive
270.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
243.65
Online exclusive
270.00 
Save 26.35 (10%)
Add to cart
Manganese(II) chloride tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number205890025
Price (USD)/ Each
243.65
Online exclusive
270.00 
Save 26.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13446-34-9
IUPAC Namemanganese(2+) tetrahydrate dichloride
Molecular FormulaCl2H8MnO4
InChI KeyCNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILESO.O.O.O.[Cl-].[Cl-].[Mn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
Sulfate (SO4)=<50 ppm
Iron (Fe)=<5 ppm
Magnesium (Mg)=<50 ppm
KMnO4-reducing substance=<0.0005 % (as O)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only