Manganese(II) sulfate monohydrate, 99+%, extra pure, Thermo Scientific Chemicals
Manganese(II) sulfate monohydrate, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, 99+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 205900050
Price (USD)/ Each
252.65
Online exclusive
281.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
252.65
Online exclusive
281.00 
Save 28.35 (10%)
Add to cart
Manganese(II) sulfate monohydrate, 99+%, extra pure, Thermo Scientific Chemicals
Catalog number205900050
Price (USD)/ Each
252.65
Online exclusive
281.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10034-96-5
IUPAC Namemanganese(2+) hydrate sulfate
Molecular FormulaH2MnO5S
InChI KeyISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILESO.[Mn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light pink
Titration Complexometric99.0 to 101.0 %
Loss on ignition10.0 % to 12.0 % (500°C)
Appearance (Form)Powder

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only