Potassium phosphate, dibasic trihydrate, 99+%, for analysis
Potassium phosphate, dibasic trihydrate, 99+%, for analysis
Potassium phosphate, dibasic trihydrate, 99+%, for analysis
Potassium phosphate, dibasic trihydrate, 99+%, for analysis
Thermo Scientific Chemicals

Potassium phosphate, dibasic trihydrate, 99+%, for analysis

CAS: 16788-57-1 | H7K2O7P | 228.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 205935000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS16788-57-1
IUPAC Namedipotassium trihydrate hydrogen phosphate
Molecular FormulaH7K2O7P
InChI KeyXQGPKZUNMMFTAL-UHFFFAOYSA-L
SMILESO.O.O.[K+].[K+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH8.5 to 9.6 (5 % solution)
Arsenic (As)=<1 ppm
Iron (Fe)=<30 ppm
Titration with HClO4>=99.0 %
Insoluble matter=<0.01 % (in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link