Thermo Scientific Chemicals

Potassium sulfate, 99+%, for analysis, anhydrous, granular, Thermo Scientific Chemicals

Catalog number: 205940025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium sulfate, 99+%, for analysis, anhydrous, granular, Thermo Scientific Chemicals

Catalog number: 205940025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 205940025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7778-80-5
IUPAC Name
dipotassium sulfate
Molecular Formula
K2O4S
InChI Key
OTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-]S([O-])(=O)=O
Appearance (Color)
White
Titration after Ion exchange
>=99.0 %
Heavy metals (as Pb)
=<5 ppm
Calcium (Ca)
=<200 ppm
Appearance (Form)
Granular powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text