Sodium nitrate, 99+%, for analysis, Thermo Scientific Chemicals
Sodium nitrate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium nitrate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Quantity:
250g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 205962500
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.60 
Save 7.95 (14%)
-
Add to cart
Quantity:
250g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.60 
Save 7.95 (14%)
Add to cart
Sodium nitrate, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number205962500
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.60 
Save 7.95 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium nitrate
Molecular FormulaNNaO3
InChI KeyVWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES[Na+].[O-][N+]([O-])=O
Molecular Weight (g/mol)84.99
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Nitrite (NO2)=<10 ppm
Iron (Fe)=<3 ppm
Appearance (Form)Crystalline powder or crystals
Heavy metals (as Pb)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only