Chemical Identifiers
IUPAC Nametetrasodium decahydrate (phosphonatooxy)phosphonate
Molecular FormulaH20Na4O17P2
InChI KeyVZWGHDYJGOMEKT-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
Molecular Weight (g/mol)446.05
View more