Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 205980050
Price (USD)/ Each
-
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7446-20-0
IUPAC Namezinc(2+) heptahydrate sulfate
Molecular FormulaH14O11SZn
InChI KeyRZLVQBNCHSJZPX-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=99.4 %
Nitrogen compounds (as N)=<0.002 %
Arsenic (As)=<1 ppm
Potassium (K)=<10 ppm
Appearance (Form)Crystalline powder or crystals and/or chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only