Thermo Scientific Chemicals

Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 205982500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 205982500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 205982500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7446-20-0
IUPAC Name
zinc(2+) heptahydrate sulfate
Molecular Formula
H14O11SZn
InChI Key
RZLVQBNCHSJZPX-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
Titration Complexometric
>=99.4 %
Nitrogen compounds (as N)
=<0.002 %
Arsenic (As)
=<1 ppm
Potassium (K)
=<10 ppm
Appearance (Form)
Crystalline powder or crystals and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text