Sodium hydroxide, 98.5%, for analysis, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 98.5%, for analysis, pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hydroxide, 98.5%, for analysis, pellets, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
10 kg
25 kg
Packaging:
View all
Plastic Bottle
Catalog number 206060025
Price (USD)/ Each
80.65
Online exclusive
89.10 
Save 8.45 (9%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
80.65
Online exclusive
89.10 
Save 8.45 (9%)
Add to cart
Sodium hydroxide, 98.5%, for analysis, pellets, Thermo Scientific Chemicals
Catalog number206060025
Price (USD)/ Each
80.65
Online exclusive
89.10 
Save 8.45 (9%)
-
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl98.0 to 102.0 %
Sodium carbonate (Na2CO3)=<0.5 %
Nitrogen compounds (as N)=<0.0003 %
Silicon dioxide (SiO2)=<20 ppm
Chloride (Cl)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only