Thermo Scientific Chemicals

1-Butanesulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 206101000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Butanesulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 206101000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 206101000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2386-54-1
IUPAC Name
sodium butane-1-sulfonate
Molecular Formula
C4H9NaO3S
InChI Key
XQCHMGAOAWZUPI-UHFFFAOYSA-M
SMILES
[Na+].CCCCS([O-])(=O)=O
Appearance (Form)
Crystalline powder
Loss on drying
=<2 % (1 g, 105°C, 2 h)
Solubility
(10 % in water) Clear to slightly hazy colorless
Appearance (Color)
White to off-white
Titration after Ion exchange
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text