Thermo Scientific Chemicals

1-hexanesulfonic acid, sodium salt hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 206120250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-hexanesulfonic acid, sodium salt hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 206120250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 206120250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2832-45-3
IUPAC Name
sodium hexane-1-sulfonate
Molecular Formula
C6H13NaO3S
InChI Key
QWSZRRAAFHGKCH-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCS([O-])(=O)=O
Appearance (Color)
White
Appearance (Form)
Powder or crystals or flakes or crystalline solid
Loss on drying
=<9.6 % (100°C, 3 h) (vacuum)
Solubility
(10 % in water) Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text