Thermo Scientific Chemicals

1-Heptanesulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 206130050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Heptanesulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 206130050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 206130050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
22767-50-6
IUPAC Name
sodium heptane-1-sulfonate
Molecular Formula
C7H15NaO3S
InChI Key
REFMEZARFCPESH-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCS([O-])(=O)=O
Solubility
(10 % in water) Clear colorless
Infrared spectrum
Conforms
Appearance (Form)
Powder or flakes
Loss on drying
=<9 % (100°C, 3 h) (vacuum)
Titration after Ion exchange
>=97.5 % (on dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text