Thermo Scientific Chemicals

1-Octanesulfonic acid, sodium salt monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 206140050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Octanesulfonic acid, sodium salt monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 206140050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 206140050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
207596-29-0
IUPAC Name
sodium hydrate octane-1-sulfonate
Molecular Formula
C8H19NaO4S
InChI Key
MBURIAHQXJQKRE-UHFFFAOYSA-M
SMILES
O.[Na+].CCCCCCCCS([O-])(=O)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Appearance (Form)
Powder
Titration after Ion exchange
>=97.5 %
Loss on drying
7 % to 9 % (140°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text