Aluminum-nickel, Raney-type alloy, powder, Al-Ni 50/50
Aluminum-nickel, Raney-type alloy, powder, Al-Ni 50/50
Aluminum-nickel, Raney-type alloy, powder, Al-Ni 50/50
Thermo Scientific Chemicals

Aluminum-nickel, Raney-type alloy, powder, Al-Ni 50/50

CAS: 12635-29-9 | AlNi | 85.675 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 206231000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12635-29-9
IUPAC Namealuminium nickel
Molecular FormulaAlNi
InChI KeyNPXOKRUENSOPAO-UHFFFAOYSA-N
SMILES[Al].[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Nickel (Ni)48.0 to 53.0 %
Aluminium (Al)47.0 to 52.0 %
Appearance (Color)Grey to grey-black
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

Is Aluminum-nickel, Raney-type Alloy, Powder Al-Ni 50/50 (Cat. No. 206231000, 206230025) supplied in weight or atomic ratio?

Aluminum-nickel, Raney-type Alloy, Powder Al-Ni 50/50 (Cat. No. 206231000, 206230025) is supplied in weight.