Thermo Scientific Chemicals

Ammonium oxalate monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 206275000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium oxalate monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 206275000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 206275000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6009-70-7
IUPAC Name
oxalic acid diamine hydrate
Molecular Formula
C2H10N2O5
InChI Key
MSMNVXKYCPHLLN-UHFFFAOYSA-N
SMILES
N.N.O.OC(=O)C(O)=O
Appearance (Color)
White
Sulfate (SO4)
=<50 ppm
Titration with KMnO4
99.4 to 101.0 %
Chloride (Cl)
=<20 ppm
Iron (Fe)
=<3 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text