Sodium tetraborate, 98%, for analysis, anhydrous, Thermo Scientific Chemicals
Sodium tetraborate, 98%, for analysis, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium tetraborate, 98%, for analysis, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 206295000
Price (USD)/ Each
66.65
Online exclusive
74.40 
Save 7.75 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
66.65
Online exclusive
74.40 
Save 7.75 (10%)
Add to cart
Sodium tetraborate, 98%, for analysis, anhydrous, Thermo Scientific Chemicals
Catalog number206295000
Price (USD)/ Each
66.65
Online exclusive
74.40 
Save 7.75 (10%)
-
Add to cart
Chemical Identifiers
CAS1330-43-4
IUPAC Namedisodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular FormulaB4Na2O7
InChI KeyNXSVMNYRVBQCMO-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<50 ppm
Phosphate (PO4)=<50 ppm
Copper (Cu)=<50 ppm
Appearance (Form)Powder or crystals
Chloride (Cl)=<500 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only