Boron oxide, 98%, -40 mesh, Thermo Scientific Chemicals
Boron oxide, 98%, -40 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron oxide, 98%, -40 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Glass Bottle
Catalog number 206300010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1303-86-2
IUPAC Nameoxo[(oxoboranyl)oxy]borane
Molecular FormulaB2O3
InChI KeyJKWMSGQKBLHBQQ-UHFFFAOYSA-N
SMILESO=BOB=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Color)White
Total trace metal impurities=<0.2 %
Impurity=<0.01 % (Total halides impurities)
Appearance (Form)Powder

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only