Thermo Scientific Chemicals

Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 206311000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 206311000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 206311000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10378-47-9
IUPAC Name
λ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
Molecular Formula
CeH20N4O18S4
InChI Key
VCNAMBGKEDPVGQ-UHFFFAOYSA-J
SMILES
N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
Appearance (Form)
Crystalline powder
Chloride (Cl)
=<100 ppm
Appearance (Color)
Yellow to orange
Titration with Na2S2O3
>=99.0 %
Iron (Fe)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text