Thermo Scientific Chemicals

Copper(II) bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 206335000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Copper(II) bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 206335000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 206335000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-45-9
IUPAC Name
copper(2+) dibromide
Molecular Formula
Br2Cu
InChI Key
QTMDXZNDVAMKGV-UHFFFAOYSA-L
SMILES
[Cu++].[Br-].[Br-]
Appearance (Color)
Dark grey to grey-black
Insoluble matter
=<0.05 % (in water)
Chloride (Cl)
=<5000 ppm
Appearance (Form)
Crystalline powder
Titration Iodometric
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text