Sodium molybdate(VI) dihydrate, 99+%
Sodium molybdate(VI) dihydrate, 99+%
Sodium molybdate(VI) dihydrate, 99+%
Sodium molybdate(VI) dihydrate, 99+%
Thermo Scientific Chemicals

Sodium molybdate(VI) dihydrate, 99+%

CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 206370050
Price (USD)
50.65
Online Exclusive
56.10
Save 5.45 (10%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
50.65
Online Exclusive
56.10
Save 5.45 (10%)
Each
Add to cart
Chemical Identifiers
CAS10102-40-6
IUPAC Namedisodium dioxomolybdenumbis(olate) dihydrate
Molecular FormulaH4MoNa2O6
InChI KeyFDEIWTXVNPKYDL-UHFFFAOYSA-N
SMILESO.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=99.0 %
Appearance (Form)Crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection