Zirconium(IV) chloride, 98%, anhydrous, Thermo Scientific Chemicals
Zirconium(IV) chloride, 98%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) chloride, 98%, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 206411000
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
Add to cart
Zirconium(IV) chloride, 98%, anhydrous, Thermo Scientific Chemicals
Catalog number206411000
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.70 
Save 11.05 (14%)
-
Add to cart
Chemical Identifiers
CAS10026-11-6
IUPAC Namezirconium(4+) tetrachloride
Molecular FormulaCl4Zr
InChI KeyDUNKXUFBGCUVQW-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Zr+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Assay>=97.5 % (ZrCl4)
Aluminium (Al)=<100 ppm
Chromium (Cr)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only