Thermo Scientific Chemicals

Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 206450010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 206450010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 206450010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7782-68-5
IUPAC Name
lithium(1+) iodate
Molecular Formula
ILiO3
InChI Key
FZAXZVHFYFGNBX-UHFFFAOYSA-M
SMILES
[Li+].[O-][I](=O)=O
Appearance (Color)
White to light beige
Titration Iodometric
>=99.4 %
Heavy metals (as Pb)
=<50 ppm
Iron (Fe)
=<30 ppm
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text