Thermo Scientific Chemicals

Ammonium iodide, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 206465000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Ammonium iodide, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 206465000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 206465000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12027-06-4
IUPAC Name
ammonium iodide
Molecular Formula
H4IN
InChI Key
UKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES
[NH4+].[I-]
pH
4.5 to 6.5 (5 % at 20°C)
Chloride and Bromide
=<0.05 % (as Cl)
Phosphate (PO4)
=<10 ppm
Calcium (Ca)
=<20 ppm
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text