Thermo Scientific Chemicals

Ammonium thiocyanate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 206500010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium thiocyanate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 206500010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 206500010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1762-95-4
IUPAC Name
ammonium cyanosulfanide
Molecular Formula
CH4N2S
InChI Key
SOIFLUNRINLCBN-UHFFFAOYSA-N
SMILES
[NH4+].[S-]C#N
Appearance (Color)
White
Water
=<1.0 %
Appearance (Form)
Crystals
Titration Argentometric
>=99.0 % (on dried basis)
pH
4.0 to 6.0 (5 % in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text